Learn More
Biomatik Corporation Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 (ENOX1), 100UG
Supplier:  Biomatik Corporation RPC21264
Encompass_Preferred
Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 (ENOX1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ENOX1. Target Synonyms: Candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase. Accession Number: Q8TC92. Expression Region: 1~643aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 77.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4990