Learn More
Biomatik Corporation Recombinant Human Tissue factor (F3), partial, 100UG
Catalog No. 501504994
Encompass_Preferred
Recombinant Human Tissue factor (F3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: F3. Target Synonyms: Coagulation factor IIIThromboplastin; CD142. Accession Number: P13726. Expression Region: 33~251aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 40.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4994 Supplier Biomatik Corporation Supplier No. RPC21292
Recombinant Human Tissue factor (F3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: F3. Target Synonyms: Coagulation factor IIIThromboplastin; CD142. Accession Number: P13726. Expression Region: 33~251aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 40.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.