Learn More
Biomatik Corporation Recombinant Bovine Guanine nucleotide-binding protein G (t) subunit alpha-2 (GNAT2), 100UG
Supplier:  Biomatik Corporation RPC21393
Encompass_Preferred
Recombinant Bovine Guanine nucleotide-binding protein G (t) subunit alpha-2 (GNAT2) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bovine (Bos taurus). Target Name: GNAT2. Target Synonyms: Transducin alpha-2 chain. Accession Number: P04696. Expression Region: 2~354aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 44kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5004