Learn More
Biomatik Corporation Recombinant Rat Glutathione S-transferase P (Gstp1), 100UG
Supplier:  Biomatik Corporation RPC21416
Encompass_Preferred
Recombinant Rat Glutathione S-transferase P (Gstp1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Gstp1. Target Synonyms: Chain 7GST 7-7GST class-pi. Accession Number: P04906. Expression Region: 2~210aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5006