Learn More
Biomatik Corporation Recombinant Human Glycophorin-A (GYPA), partial, 100UG
Catalog No. 501505007
Encompass_Preferred
Recombinant Human Glycophorin-A (GYPA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GYPA. Target Synonyms: MN sialoglycoprotein; PAS-2Sialoglycoprotein alpha; CD235a. Accession Number: A0A0C4DFT7. Expression Region: 20~91aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 34.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5007 Supplier Biomatik Corporation Supplier No. RPC21424
Recombinant Human Glycophorin-A (GYPA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GYPA. Target Synonyms: MN sialoglycoprotein; PAS-2Sialoglycoprotein alpha; CD235a. Accession Number: A0A0C4DFT7. Expression Region: 20~91aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 34.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.