Learn More
Biomatik Corporation Recombinant Larimichthys crocea Hepcidin (hamp), 100UG
Catalog No. 501505008
Encompass_Preferred
Recombinant Larimichthys crocea Hepcidin (hamp) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Larimichthys crocea (Large yellow croaker) (Pseudosciaena crocea). Target Name: hamp. Target Synonyms: hamp; Hepcidin. Accession Number: A1Z0M0. Expression Region: 65~85aa. Tag Info: Tag-Free. Theoretical MW: 2.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5008 Supplier Biomatik Corporation Supplier No. RPC21433
Recombinant Larimichthys crocea Hepcidin (hamp) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Larimichthys crocea (Large yellow croaker) (Pseudosciaena crocea). Target Name: hamp. Target Synonyms: hamp; Hepcidin. Accession Number: A1Z0M0. Expression Region: 65~85aa. Tag Info: Tag-Free. Theoretical MW: 2.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.