Learn More
Biomatik Corporation Recombinant Mouse 3-hydroxy-3-methylglutaryl-coenzyme A reductase (Hmgcr), partial, 100UG
Catalog No. 501505010
Encompass_Preferred
Recombinant Mouse 3-hydroxy-3-methylglutaryl-coenzyme A reductase (Hmgcr), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Hmgcr. Target Synonyms: Hmgcr; 3-hydroxy-3-methylglutaryl-coenzyme A reductase; HMG-CoA reductase; EC 1.1.1.34. Accession Number: Q01237. Expression Region: 700~860aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 20.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5010 Supplier Biomatik Corporation Supplier No. RPC21459
Recombinant Mouse 3-hydroxy-3-methylglutaryl-coenzyme A reductase (Hmgcr), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Hmgcr. Target Synonyms: Hmgcr; 3-hydroxy-3-methylglutaryl-coenzyme A reductase; HMG-CoA reductase; EC 1.1.1.34. Accession Number: Q01237. Expression Region: 700~860aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 20.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.