Learn More
Biomatik Corporation Recombinant Mouse Protein S100-A8 (S100a8), 100UG
Supplier:  Biomatik Corporation RPC20078
Encompass_Preferred
Recombinant Mouse Protein S100-A8 (S100a8) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: S100a8. Target Synonyms: Calgranulin-AChemotactic cytokine CP-10Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; p8Pro-inflammatory S100 cytokine; S100 calcium-binding protein A8. Accession Number: P27005. Expression Region: 2~89aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 37.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4578