Learn More
Biomatik Corporation RecombinantHumanKeratin,typeII cytoskeletal 8 (KRT8), partial, 100UG
Supplier:  Biomatik Corporation RPC21583
Encompass_Preferred
Recombinant Human Keratin, type II cytoskeletal 8 (KRT8), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: KRT8. Target Synonyms: Cytokeratin-8; CK-8Keratin-8; K8Type-II keratin Kb8. Accession Number: P05787. Expression Region: 2~483aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 66.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5020