Learn More
Biomatik Corporation Recombinant Human Laminin subunit alpha-5 (LAMA5), partial, 100UG
Supplier:  Biomatik Corporation RPC21586
Encompass_Preferred
Recombinant Human Laminin subunit alpha-5 (LAMA5), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LAMA5. Target Synonyms: Laminin-10 subunit alphaLaminin-11 subunit alphaLaminin-15 subunit alpha. Accession Number: O15230. Expression Region: 3401~3692aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 47.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5021