Learn More
Biomatik Corporation Recombinant Mouse Legumain (Lgmn), 100UG
Supplier:  Biomatik Corporation RPC21602
Encompass_Preferred
Recombinant Mouse Legumain (Lgmn) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Lgmn. Target Synonyms: Asparaginyl endopeptidaseProtease, cysteine 1. Accession Number: O89017. Expression Region: 18~325aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 38.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5024