Learn More
Biomatik Corporation Recombinant Human Protein-lysine 6-oxidase (LOX), 100UG
Catalog No. 501505025
Encompass_Preferred
Recombinant Human Protein-lysine 6-oxidase (LOX) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LOX. Target Synonyms: Lysyl oxidase. Accession Number: P28300. Expression Region: 169~417aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 49kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5025 Supplier Biomatik Corporation Supplier No. RPC21615
Recombinant Human Protein-lysine 6-oxidase (LOX) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LOX. Target Synonyms: Lysyl oxidase. Accession Number: P28300. Expression Region: 169~417aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 49kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.