Learn More
Biomatik Corporation Recombinant Mouse Lymphotoxin-alpha (Lta), 100UG
Supplier:  Biomatik Corporation RPC21627
Encompass_Preferred
Recombinant Mouse Lymphotoxin-alpha (Lta) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Lta. Target Synonyms: TNF-betaTumor necrosis factor ligand superfamily member 1Tnfb, Tnfsf1. Accession Number: P09225. Expression Region: 34~202aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 23.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5026