Learn More
Biomatik Corporation Recombinant Human Lymphocyte antigen 6 complex locus protein G6d (LY6G6D), partial, 100UG
Supplier:  Biomatik Corporation RPC21630
Encompass_Preferred
Recombinant Human Lymphocyte antigen 6 complex locus protein G6d (LY6G6D), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LY6G6D. Target Synonyms: Megakaryocyte-enhanced gene transcript 1 protein. Accession Number: O95868. Expression Region: 10~104aa. Tag Info: N-Terminal 10Xhis-B2M-Jd-Tagged. Theoretical MW: 15.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5028