Learn More
Biomatik Corporation Recombinant Rat Protein S100-A8 (S100a8), 100UG
Catalog No. 501504579
Encompass_Preferred
Recombinant Rat Protein S100-A8 (S100a8) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: S100a8. Target Synonyms: Calgranulin-AMigration inhibitory factor-related protein 8; MRP-8; p8S100 calcium-binding protein A8. Accession Number: P50115. Expression Region: 2~89aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 14.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4579 Supplier Biomatik Corporation Supplier No. RPC20079
Recombinant Rat Protein S100-A8 (S100a8) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: S100a8. Target Synonyms: Calgranulin-AMigration inhibitory factor-related protein 8; MRP-8; p8S100 calcium-binding protein A8. Accession Number: P50115. Expression Region: 2~89aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 14.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.