Learn More
Biomatik Corporation Recombinant Human Melanoma-associated antigen 10 (MAGEA10), 100UG
Catalog No. 501505030
Encompass_Preferred
Recombinant Human Melanoma-associated antigen 10 (MAGEA10) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MAGEA10. Target Synonyms: Cancer/testis antigen 1.10. Accession Number: P43363. Expression Region: 1~369aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 46.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5030 Supplier Biomatik Corporation Supplier No. RPC21638
Recombinant Human Melanoma-associated antigen 10 (MAGEA10) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MAGEA10. Target Synonyms: Cancer/testis antigen 1.10. Accession Number: P43363. Expression Region: 1~369aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 46.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.