Learn More
Biomatik Corporation Recombinant Human Methylmalonyl-CoA mutase, mitochondrial (MUT), 100UG
Supplier:  Biomatik Corporation RPC21704
Encompass_Preferred
Recombinant Human Methylmalonyl-CoA mutase, mitochondrial (MMUT) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MMUT. Target Synonyms: Methylmalonyl-CoA isomerase. Accession Number: P22033. Expression Region: 33~750aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 84.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5036