Learn More
Biomatik Corporation Recombinant Rat Neural cell adhesion molecule 1 (Ncam1), partial, 100UG
Supplier:  Biomatik Corporation RPC21718
Encompass_Preferred
Recombinant Rat Neural cell adhesion molecule 1 (Ncam1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Ncam1. Target Synonyms: CD_antigen: CD56. Accession Number: P13596. Expression Region: 20~721aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 82.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5038