Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Acyl carrier protein (acpP), 100UG
Supplier:  Biomatik Corporation RPC21724
Encompass_Preferred
Recombinant Staphylococcus aureus Acyl carrier protein (acpP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain Mu50 / ATCC 700699). Target Name: acpP. Target Synonyms: hmrB. Accession Number: P0A001. Expression Region: 1~77aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 24.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5040