Learn More
Biomatik Corporation Recombinant Dog Serum amyloid A protein (SAA1), 100UG
Catalog No. 501504581
Encompass_Preferred
Recombinant Dog Serum amyloid A protein (SAA1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Dog (Canis familiaris; Canine). Target Name: SAA1. Target Synonyms: Amyloid fibril protein AA. Accession Number: P19708. Expression Region: 19~129aa. Tag Info: N-Terminal 6Xhis-Gst-Tagged. Theoretical MW: 42.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4581 Supplier Biomatik Corporation Supplier No. RPC20081
Recombinant Dog Serum amyloid A protein (SAA1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Dog (Canis familiaris; Canine). Target Name: SAA1. Target Synonyms: Amyloid fibril protein AA. Accession Number: P19708. Expression Region: 19~129aa. Tag Info: N-Terminal 6Xhis-Gst-Tagged. Theoretical MW: 42.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.