Learn More
Biomatik Corporation Recombinant Human Oxytocin-neurophysin 1 (OXT), partial, 100UG
Supplier:  Biomatik Corporation RPC21779
Encompass_Preferred
Recombinant Human Oxytocin-neurophysin 1 (OXT), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: OxT. Target Synonyms: Ocytocin. Accession Number: P01178. Expression Region: 32~125aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 14.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5049