Learn More
Biomatik Corporation Recombinant Human Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial (SDHA), partial, 100UG
Supplier:  Biomatik Corporation RPC20083
Encompass_Preferred
Recombinant Human Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial (SDHA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SDHA. Target Synonyms: Flavoprotein subunit of complex II; Fp. Accession Number: P31040. Expression Region: 44~664aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 84kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4582