Learn More
Biomatik Corporation Recombinant Human Paraneoplastic antigen Ma1 (PNMA1), 100UG
Supplier:  Biomatik Corporation RPC21835
Encompass_Preferred
Recombinant Human Paraneoplastic antigen Ma1 (PNMA1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PNMA1. Target Synonyms: 37 kDa neuronal proteinNeuron- and testis-specific protein 1. Accession Number: Q8ND90. Expression Region: 1~353aa. Tag Info: N-Terminal 10Xhis-B2M-Jd-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 46.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5059