Learn More
Biomatik Corporation Recombinant Human Serum paraoxonase/arylesterase 1 (PON1), 100UG
Catalog No. 501505061
Encompass_Preferred
Recombinant Human Serum paraoxonase/arylesterase 1 (PON1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PON1. Target Synonyms: Aromatic esterase 1; A-esterase 1K-45Serum aryldialkylphosphatase 1. Accession Number: P27169. Expression Region: 2~355aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 66.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5061 Supplier Biomatik Corporation Supplier No. RPC21840
Recombinant Human Serum paraoxonase/arylesterase 1 (PON1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PON1. Target Synonyms: Aromatic esterase 1; A-esterase 1K-45Serum aryldialkylphosphatase 1. Accession Number: P27169. Expression Region: 2~355aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 66.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.