Learn More
Biomatik Corporation Recombinant Mouse Peroxisome proliferator-activated receptor gamma (Pparg), 100UG
Supplier:  Biomatik Corporation RPC21846
Encompass_Preferred
Recombinant Mouse Peroxisome proliferator-activated receptor gamma (Pparg) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Pparg. Target Synonyms: Pparg; Nr1c3; Peroxisome proliferator-activated receptor gamma; PPAR-gamma; Nuclear receptor subfamily 1 group C member 3. Accession Number: P37238. Expression Region: 1~505aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 63.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5062