Learn More
Biomatik Corporation Recombinant Human Peroxiredoxin-1 (PRDX1), 100UG
Supplier:  Biomatik Corporation RPC21853
Encompass_Preferred
Recombinant Human Peroxiredoxin-1 (PRDX1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PRDX1. Target Synonyms: Natural killer cell-enhancing factor AProliferation-associated gene proteinThioredoxin peroxidase 2Thioredoxin-dependent peroxide reductase 2PAGA, PAGB, TDPX2. Accession Number: Q06830. Expression Region: 1~199aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5063