Learn More
Biomatik Corporation Recombinant Human Vitamin K-dependent protein S (PROS1), 100UG
Supplier:  Biomatik Corporation RPC21861
Encompass_Preferred
Recombinant Human Vitamin K-dependent protein S (PROS1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PROS1. Target Synonyms: Preproprotein S; Propiece of latent protein S; PROS 1; PROS; PROS_HUMAN; proS1; Protein S alpha; Protein Sa; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS21; PS22; PS23; PS24; PS25; PS26; PSA; THPH5; THPH6; Vitamin K dependent protein S; Vitamin K-dependent plasma protein S. Accession Number: P07225. Expression Region: 42~676aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 86.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5064