Learn More
Biomatik Corporation Recombinant Human Proteasome subunit beta type-8 (PSMB8), 100UG
Catalog No. 501505066
Encompass_Preferred
Recombinant Human Proteasome subunit beta type-8 (PSMB8) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PSMB8. Target Synonyms: Low molecular mass protein 7Macropain subunit C13Multicatalytic endopeptidase complex subunit C13Proteasome component C13Proteasome subunit beta-5iReally interesting new gene 10 protein. Accession Number: P28062. Expression Region: 73~276aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 42.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5066 Supplier Biomatik Corporation Supplier No. RPC21873
Recombinant Human Proteasome subunit beta type-8 (PSMB8) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PSMB8. Target Synonyms: Low molecular mass protein 7Macropain subunit C13Multicatalytic endopeptidase complex subunit C13Proteasome component C13Proteasome subunit beta-5iReally interesting new gene 10 protein. Accession Number: P28062. Expression Region: 73~276aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 42.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.