Learn More
Biomatik Corporation Recombinant Human Selenoprotein P (SEPP1), 100UG
Supplier:  Biomatik Corporation RPC20084
Encompass_Preferred
Recombinant Human Selenoprotein P (SEPP1) (U59S, U300S, U318S, U330S, U345S, U352S, U367S, U369S, U376S, U378S) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SEPP1. Target Synonyms: Selenoprotein P; Selenoprotein P plasma 1; Selp; SeP; Sepp1; SEPP1_HUMAN. Accession Number: P49908. Expression Region: 20~381aa(U59S,U300S,U318S,U330S,U345S,U352S,U367S,U369S,U376S,U378S). Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 44.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4583