Learn More
Biomatik Corporation Recombinant Bovine Protein S100-A9 (S100A9), 100UG
Supplier:  Biomatik Corporation RPC21965
Encompass_Preferred
Recombinant Bovine Protein S100-A9 (S100A9) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bovine (Bos taurus). Target Name: S100A9. Target Synonyms: BEE22Calgranulin-BNeutrophil cytosolic 23 kDa protein. Accession Number: P28783. Expression Region: 2~156aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 34.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5070