Learn More
Biomatik Corporation Recombinant Mouse SLAM family member 7 (Slamf7), partial, 100UG
Supplier:  Biomatik Corporation RPC22008
Encompass_Preferred
Recombinant Mouse SLAM family member 7 (Slamf7), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Slamf7. Target Synonyms: Leukocyte cell-surface antigenNovel Ly9CD_antigen: CD319. Accession Number: Q8BHK6. Expression Region: 23~224aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5072