Learn More
Biomatik Corporation Recombinant Human Extracellular superoxide dismutase [Cu-Zn] (SOD3), 100UG
Catalog No. 501505078
Encompass_Preferred
Recombinant Human Extracellular superoxide dismutase [Cu-Zn] (SOD3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SOD3. Target Synonyms: EC SOD; EC-SOD; Extracellular superoxide dismutase [Cu Zn]; Extracellular superoxide dismutase [Cu-Zn]; Extracellular superoxide dismutase; Extracellular superoxide dismutase precursor; MGC20077; SOD 3; SOD3; SODE_HUMAN; Superoxide Dismutase 3. Accession Number: P08294. Expression Region: 19~240aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5078 Supplier Biomatik Corporation Supplier No. RPC22039
Recombinant Human Extracellular superoxide dismutase [Cu-Zn] (SOD3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SOD3. Target Synonyms: EC SOD; EC-SOD; Extracellular superoxide dismutase [Cu Zn]; Extracellular superoxide dismutase [Cu-Zn]; Extracellular superoxide dismutase; Extracellular superoxide dismutase precursor; MGC20077; SOD 3; SOD3; SODE_HUMAN; Superoxide Dismutase 3. Accession Number: P08294. Expression Region: 19~240aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.