Learn More
Biomatik Corporation Recombinant Human Hyaluronidase PH-20 (SPAM1), 100UG
Supplier:  Biomatik Corporation RPC22044
Encompass_Preferred
Recombinant Human Hyaluronidase PH-20 (SPAM1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SPAM1. Target Synonyms: Hyaluronoglucosaminidase PH-20Sperm adhesion molecule 1Sperm surface protein PH-20. Accession Number: P38567. Expression Region: 36~490aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 67.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5080