Learn More
Biomatik Corporation Recombinant Rat Sulfotransferase family cytosolic 1B member 1 (Sult1b1), 100UG
Supplier:  Biomatik Corporation RPC22076
Encompass_Preferred
Recombinant Rat Sulfotransferase family cytosolic 1B member 1 (Sult1b1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Sult1b1. Target Synonyms: DOPA/tyrosine sulfotransferase. Accession Number: P52847. Expression Region: 1~299aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 54.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5085