Learn More
Biomatik Corporation Recombinant Rat Alpha-synuclein (Snca), 100UG
Catalog No. 501504585
Encompass_Preferred
Recombinant Rat Alpha-synuclein (Snca) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Snca. Target Synonyms: Snca; Alpha-synuclein. Accession Number: P37377. Expression Region: 1~140aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 41.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4585 Supplier Biomatik Corporation Supplier No. RPC20087
Recombinant Rat Alpha-synuclein (Snca) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Snca. Target Synonyms: Snca; Alpha-synuclein. Accession Number: P37377. Expression Region: 1~140aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 41.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.