Learn More
Biomatik Corporation Recombinant Mouse Tryptase beta-2 (Tpsb2), 100UG
Supplier:  Biomatik Corporation RPC22166
Encompass_Preferred
Recombinant Mouse Tryptase beta-2 (Tpsb2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Tpsb2. Target Synonyms: Mast cell protease 6; mMCP-6. Accession Number: P21845. Expression Region: 32~276aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 54.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5093