Learn More
Biomatik Corporation Recombinant Mouse Transthyretin (Ttr), partial, 100UG
Supplier:  Biomatik Corporation RPC22186
Encompass_Preferred
Recombinant Mouse Transthyretin (Ttr), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ttr. Target Synonyms: Prealbumin. Accession Number: P07309. Expression Region: 23~147aa. Tag Info: Tag-Free. Theoretical MW: 13.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5098