Learn More
Biomatik Corporation Recombinant Human Thioredoxin domain-containing protein 17 (TXNDC17), 100UG
Supplier:  Biomatik Corporation RPC22195
Encompass_Preferred
Recombinant Human Thioredoxin domain-containing protein 17 (TXNDC17) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TXNDC17. Target Synonyms: 14 kDa thioredoxin-related protein; TRP14Protein 42-9-9Thioredoxin-like protein 5. Accession Number: Q9BRA2. Expression Region: 1~123aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 40.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5101