Learn More
Biomatik Corporation Recombinant Human UDP-glucose 6-dehydrogenase (UGDH), 100UG
Supplier:  Biomatik Corporation RPC22208
Encompass_Preferred
Recombinant Human UDP-glucose 6-dehydrogenase (UGDH) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: UGDH. Target Synonyms: GDH; UDP Glc dehydrogenase; UDP GlcDH; UDP glucose 6 dehydrogenase; UDP glucose dehydrogenase; UDP-Glc dehydrogenase; UDP-GlcDH; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogenase; UDPGDH; UGD; Ugdh; UGDH_HUMAN; Uridine diphospho glucose dehydrogenase. Accession Number: O60701. Expression Region: 1~494aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 71kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5102