Learn More
Biomatik Corporation Recombinant Human Tryptophan--tRNA ligase, cytoplasmic (WARS), 100UG
Supplier:  Biomatik Corporation RPC22226
Encompass_Preferred
Recombinant Human Tryptophan--tRNA ligase, cytoplasmic (WARS) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: WARS. Target Synonyms: Interferon-induced protein 53Tryptophanyl-tRNA synthetaseIFI53, WRS. Accession Number: P23381. Expression Region: 2~471aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 60kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5105