Learn More
Biomatik Corporation Recombinant Human Protein Wnt-4 (WNT4), 100UG
Supplier:  Biomatik Corporation RPC22234
Encompass_Preferred
Recombinant Human Protein Wnt-4 (WNT4) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: WNT4. Target Synonyms: MGC123964; Protein Wnt-4; RP23-246F18.1; SERKAL; Wingless type MMTV integration site family member 4 precursor; WNT 4; WNT 4 protein precursor; WNT4; WNT4_HUMAN. Accession Number: P56705. Expression Region: 23~351aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 52.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5106