Learn More
Biomatik Corporation Recombinant Rat Sortilin (Sort1), partial, 100UG
Catalog No. 501504587
Encompass_Preferred
Recombinant Rat Sortilin (Sort1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Sort1. Target Synonyms: Glycoprotein 110; Gp110Neurotensin receptor 3; NTR3. Accession Number: O54861. Expression Region: 610~754aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 32.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4587 Supplier Biomatik Corporation Supplier No. RPC20089
Recombinant Rat Sortilin (Sort1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Sort1. Target Synonyms: Glycoprotein 110; Gp110Neurotensin receptor 3; NTR3. Accession Number: O54861. Expression Region: 610~754aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 32.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.