Learn More
Biomatik Corporation Recombinant Plasmodium berghei L-lactate dehydrogenase (PB000185.00.0), 100UG
Supplier:  Biomatik Corporation RPC22346
Encompass_Preferred
Recombinant Plasmodium berghei L-lactate dehydrogenase (PB000185.00.0) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Plasmodium berghei (strain Anka). Target Name: PB000185.00.0. Target Synonyms: PB000185.00.0; L-lactate dehydrogenase; EC 1.1.1.27. Accession Number: Q7SI97. Expression Region: 1~316aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 39.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5114