Learn More
Biomatik Corporation Recombinant Avena sativa Endochitinase, 100UG
Supplier:  Biomatik Corporation RPC22355
Encompass_Preferred
Recombinant Avena sativa Endochitinase is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Avena sativa (Oat). Target Name: Avena sativa Endochitinase. Target Synonyms: Endochitinase; EC 3.2.1.14; Fragments. Accession Number: P86181. Expression Region: 1~200aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 25.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5115