Learn More
Biomatik Corporation Recombinant Rhesus cytomegalovirus Envelope glycoprotein B (gB), partial, 100UG
Supplier:  Biomatik Corporation RPC22360
Encompass_Preferred
Recombinant Rhesus cytomegalovirus Envelope glycoprotein B (gB), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rhesus cytomegalovirus (strain 68-1) (RhCMV). Target Name: gB. Target Synonyms: gB; UL55; Envelope glycoprotein B; gB. Accession Number: P89053. Expression Region: 745~854aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 18kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5116