Learn More
Biomatik Corporation Recombinant Pseudomonas aeruginosa Outer membrane porin F (oprF), 100UG
Supplier:  Biomatik Corporation RPC22401
Encompass_Preferred
Recombinant Pseudomonas aeruginosa Outer membrane porin F (oprF) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Pseudomonas aeruginosa (strain ATCC 15692 / DSM 22644 / CIP 104116 / JCM 14847 / LMG 12228 / 1C / PRS 101 / PAO1). Target Name: oprF. Target Synonyms: oprF; PA1777; Outer membrane porin F. Accession Number: P13794. Expression Region: 25~350aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 39.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5119