Learn More
Biomatik Corporation Recombinant Streptomyces alboniger Puromycin N-acetyltransferase (pac), 100UG
Catalog No. 501505120
Encompass_Preferred
Recombinant Streptomyces alboniger Puromycin N-acetyltransferase (pac) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Streptomyces alboniger. Target Name: pac. Target Synonyms: pac; Puromycin N-acetyltransferase; EC 2.3. Accession Number: P13249. Expression Region: 1~199aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 25.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5120 Supplier Biomatik Corporation Supplier No. RPC22414
Recombinant Streptomyces alboniger Puromycin N-acetyltransferase (pac) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Streptomyces alboniger. Target Name: pac. Target Synonyms: pac; Puromycin N-acetyltransferase; EC 2.3. Accession Number: P13249. Expression Region: 1~199aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 25.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.