Learn More
Biomatik Corporation Recombinant Synsepalum dulcificum Miraculin, 100UG
Supplier:  Biomatik Corporation RPC22421
Encompass_Preferred
Recombinant Synsepalum dulcificum Miraculin is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Synsepalum dulcificum (Miracle fruit) (Richadella dulcifica). Target Name: Synsepalum dulcificum Miraculin. Target Synonyms: Miraculin; MIR. Accession Number: P13087. Expression Region: 30~220aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 24.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5122