Learn More
Biomatik Corporation Recombinant Zaire ebolavirus Nucleoprotein (NP), partial, 100UG
Catalog No. 501505128
Encompass_Preferred
Recombinant Zaire ebolavirus Nucleoprotein (NP), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Zaire ebolavirus (strain Mayinga-76) (ZEBOV) (Zaire Ebola virus). Target Name: NP. Target Synonyms: Nucleocapsid protein; Protein N. Accession Number: P18272. Expression Region: 488~739aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 33.1 kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5128 Supplier Biomatik Corporation Supplier No. RPC22461
Recombinant Zaire ebolavirus Nucleoprotein (NP), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Zaire ebolavirus (strain Mayinga-76) (ZEBOV) (Zaire Ebola virus). Target Name: NP. Target Synonyms: Nucleocapsid protein; Protein N. Accession Number: P18272. Expression Region: 488~739aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 33.1 kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.