Learn More
Biomatik Corporation Recombinant Bovine Transforming growth factor beta-1 (TGFB1), partial, 100UG
Catalog No. 501504589
Encompass_Preferred
Recombinant Bovine Transforming growth factor beta-1 proprotein (TGFB1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bovine (Bos taurus). Target Name: TGFB1. Target Synonyms: TGFB1; Transforming growth factor beta-1 proprotein; Cleaved into: Latency-associated peptide; LAP; Transforming growth factor beta-1; TGF-beta-1. Accession Number: P18341. Expression Region: 279~390aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 16.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4589 Supplier Biomatik Corporation Supplier No. RPC20094
Recombinant Bovine Transforming growth factor beta-1 proprotein (TGFB1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bovine (Bos taurus). Target Name: TGFB1. Target Synonyms: TGFB1; Transforming growth factor beta-1 proprotein; Cleaved into: Latency-associated peptide; LAP; Transforming growth factor beta-1; TGF-beta-1. Accession Number: P18341. Expression Region: 279~390aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 16.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.